Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant IL33 (from E. coli)
Human Recombinant IL33 (from E. coli)
Catalog # 75793-600
Supplier:  Prosci
CAS Number:  
Human Recombinant IL33 (from E. coli)
Catalog # 75793-600
Supplier:  Prosci
Supplier Number:  90-047
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    10 μg
  • Tag sequence:
    His Tag
  • Storage Conditions:
    Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Biological Activity:
    Activates mouse and human ST2-dependent NF-kappaB pathway.
  • Protein Synonyms:
    IL-1F11|NF-HEV|Interleukin-33
  • Protein/Peptide Name:
    IL33
  • Purity:
    >90% (SDS-PAGE)
  • Sequence:
    109-266
  • Formulation:
    0.2um-filtered solution in 55mM TRIS-Cl, pH 8.2, containing 150mM NaCl.
  • Cat. No.:
    75793-600
  • Supplier no.:
    90-047

Specifications

About this item

Interleukin-33 (IL-33; HF-NEV; IL-1F11), a member of the IL-1 family of cytokines, is expressed by many cell types following pro-inflammatory stimulation and is thought to be released on cell lysis. The 30kDa human IL33 is converted by CASP1 to a 18kDa protein. IL33 binds to and signals through ST2 (IL1R1) and its stimulation recruits MYD88, IRAK, IRAK4, and TRAF6, followed by phosphorylation of ERK1 (MAPK3)/ERK2 (MAPK1), p38 (MAPK14), and JNK. The ability of IL-33 to target numerous immune cell types, like Th2-like cells, mast cells, and B1 cells, and to induce cytokine and chemokine production underlines its potential in influencing the outcome of a wide range of diseases, such as arthritis, asthma, atopic allergy & anaphylaxis, cardiovascular disease/atherosclerosis, nervous system diseases, and sepsis.

  • Fusion-Tag: His Tag

This recombinant proteins is for research use only.