Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant IL22 (from Cells)
Catalog # 75791-886
Supplier:  Prosci
CAS Number:  
Human Recombinant IL22 (from Cells)
Catalog # 75791-886
Supplier:  Prosci
Supplier Number:  92-662
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-hIgG4 Fc tag
  • Storage Conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein Synonyms:
    IL-TIF|IL-22|Cytokine Zcyto18|IL-10-related T-cell-derived-inducible factor|IL22|Interleukin-22
  • Protein/Peptide Name:
    IL22
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    43.4 kD
  • Sequence:
    Ala34-Ile179
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Cat. No.:
    75791-886
  • Supplier no.:
    92-662

Specifications

About this item

Interleukin-22(IL-22) is a member of a group of the IL-10 family, a class of potent mediators of cellular inflammatory responses. IL-22 is produced by activated DC and T cells. IL-22 and IL-10 receptor chains play a role in cellular targeting and signal transduction. It can initiate and regulate innate immune responses against bacterial pathogens especially in epithelial cells such as respiratory and gut epithelial cells. IL-22 along with IL-17 likely plays a role in the coordinated response of both adaptive and innate immune systems. IL-22 also promotes hepatocyte survival in the liver and epithelial cells in the lung and gut similar to IL-10. Biological activity of IL-22 is initiated by binding to a cell-surface complex consisting of IL-22R1 and IL-10R2 receptor chains. IL-22 biological activity is further regulated by interactions with a soluble binding protein, IL-22BP. IL-22BP and an extracellular region of IL-22R1 share sequence similarity. In some cases, the pro-inflammatory versus tissue-protective functions of IL-22 are regulated by cytokine IL-17A.

  • Fusion-Tag: C-hIgG4 Fc tag

This recombinant protein can be used for biological assays. For research use only.