Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant TIM3
Mouse Recombinant TIM3
Catalog # 75791-754
Supplier:  Prosci
CAS Number:  
Mouse Recombinant TIM3
Catalog # 75791-754
Supplier:  Prosci
Supplier Number:  92-596
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
  • Species:
    Mouse
  • Size:
    0.05 mg
  • Environmentally Preferable:
  • Tag sequence:
    C-Fc tag
  • Storage Conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein Synonyms:
    Hepatitis A virus cellular receptor 2 homolog|T-cell membrane protein 3|HAVcr-2|T-cell immunoglobulin and mucin domain-containing protein 3|Timd3|T-cell immunoglobulin mucin receptor 3|Tim3|HAVCR2
  • Protein/Peptide Name:
    TIM3
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    46.3 kD
  • Sequence:
    Arg20-Arg191
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Cat. No.:
    75791-754
  • Supplier no.:
    92-596

Specifications

About this item

T cell immunoglobulin and mucin domain-3 (TIM3), also called hepatitis A virus cellular receptor 2 (HAVCR2), is a transmembrane glycoprotein of the TIM family of immune regulating molecules and plays an important role in the Th1-mediated immune response. TIM3 is expressed on the Th1 cells, CD8 T-cells, monocytes, and dendritic cells, but not on Th2 cells. TIM3 expressed by monocytes and dendritic cells facilitates phagocytosis of apoptotic cells and up-regulates cross-presentation of apoptotic cell-associated antigens through interaction with phosphatidylserine. Engagement of TIM3 by its ligand galectin-9 induces a range of immunosuppressive functions which enhance immune tolerance and inhibit anti-tumor immunity. Stimulation of TIM3 with an agonistic antibody promotes inflammation through the activation of innate immune cells. TIM3 is also regarded as a potential target molecule for immunotherapy. TIM3 and programmed cell death 1 (PD-1) as two important coinhibitory regulators of T cell responses, have been implicated with the T-cell dysfunction or exhaustion associated with chronic HBV infection including HBV-related HCC.

  • Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.