Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Peroxiredoxin 5 (from Cells)
Human Recombinant Peroxiredoxin 5 (from Cells)
Catalog # 75791-732
Supplier:  Prosci
CAS Number:  
Human Recombinant Peroxiredoxin 5 (from Cells)
Catalog # 75791-732
Supplier:  Prosci
Supplier Number:  92-585
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    N-6 His tag
  • Storage Conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein Synonyms:
    Liver tissue 2D-page spot 71B|Antioxidant enzyme B166|Peroxisomal antioxidant enzyme|TPx type VI|PRDX5|AOEB166|Alu corepressor 1|Thioredoxin reductase|Prx-V|Peroxiredoxin V|Thioredoxin peroxidase PMP20|Peroxiredoxin-5|PLP
  • Protein/Peptide Name:
    Peroxiredoxin 5
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    17.9 kD
  • Sequence:
    Met53-Leu214
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Cat. No.:
    75791-732
  • Supplier no.:
    92-585

Specifications

About this item

Peroxisomes are essential organelles that participate in multiple important metabolic processes, including the beta -oxidation of fatty acids, plasmalogen synthesis, and the metabolism of reactive oxygen species (ROS). Peroxiredoxins is overexpressed in breast cancer tissues to a great extent suggesting that they has a proliferative effect and may be related to cancer development or progression. Peroxiredoxin 5 (PRDX5) is a thioredoxin peroxidase that belongs to the atypical 2-Cys class of the TSA/ahpC family of peroxiredoxins. PRDX5 is a widely expressed mitochondrial antioxidant enzyme that reduces hydrogen peroxide, alkyl hydroperoxides, and peroxynitrite. In human cells, this enzyme is present in the cytosol, mitochondria, peroxisomes, and nucleus.

  • Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.