Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant ALK1
Mouse Recombinant ALK1
Catalog # 75791-692
Supplier:  Prosci
CAS Number:  
Mouse Recombinant ALK1
Catalog # 75791-692
Supplier:  Prosci
Supplier Number:  92-565
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Species:
    Mouse
  • Size:
    0.05 mg
  • Tag sequence:
    C-Fc tag
  • Storage Conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein Synonyms:
    activin A receptor|Activin receptor-like kinase 1|ACVRL1|SKR3|ALK-1|activin A receptor type II-like 1|TSR-I|Serine/threonine-protein kinase receptor R3|TGF-B superfamily receptor type I
  • Protein/Peptide Name:
    ALK1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    38.1 kD
  • Sequence:
    Asp23-Pro119
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Cat. No.:
    75791-692
  • Supplier no.:
    92-565

Specifications

About this item

Activin Receptor-Like Kinase 1 (ALK-1) is a type I cell-surface receptor for the TGF- beta superfamily of ligands, which mediates signaling of BMP9 (bone morphogenetic protein) and BMP10. ALK1 signaling is necessary for angiogenesis during embryogenesis, wound healing, and tumor growth. ALK-1 has a high degree of similarity in serine-threonine kinase subdomains, a glycine and serine rich region preceding the kinase-domain, and a C-terminal tail with other activin receptor-like kinase proteins. ALK-1 is mainly expressed in endothelial cells regulating proliferation and migration in vitro and angiogenesis in vivo. Mutations in ALK-1 as well as in endoglin are associated with hereditary hemorrhagic telangiectasia (HHT), suggesting ALK-1 plays a critical role for in the control of blood vessel development or repair.

  • Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.