Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant Chordin-like 2
Mouse Recombinant Chordin-like 2
Catalog # 75791-690
Supplier:  Prosci
CAS Number:  
Mouse Recombinant Chordin-like 2
Catalog # 75791-690
Supplier:  Prosci
Supplier Number:  92-564
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Species:
    Mouse
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage Conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein Synonyms:
    BNF-1|CHL2|chordin-like 2|Chordin-like protein 2|CHRDL2
  • Protein/Peptide Name:
    Chordin-like 2
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    46.3 kD
  • Sequence:
    Gln24-Leu426
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Cat. No.:
    75791-690
  • Supplier no.:
    92-564

Specifications

About this item

Mouse Chordin-Like 2, also known as CHL2, is a novel chordin family member with structural homology to CHL1 which is implicated in tumor angiogenesis. The mouse CHL2 gene encodes a 426 amino acids (aa) protein with a 25 aa signal peptide. The mature chain of CHL2 protein contains two potential N-linked glycosylation sites, one putative NLS and three 63 aa cysteine-rich von Willebrand type C repeats (CRs). CHL2 gene is weakly expressed in the liver and kidney, partly expressed in the connective tissues of reproductive organs such as ligaments of the ovary and oviduct in females, and of testis, epididymis and certain male accessory sex glands in males. Recombinant mCHL2 protein interacted directly with five BMPs and one GDF thereby inhibiting, in vitro, several BMP/GDF-dependent processes including, osteogenic differentiation of C2C12 mesenchymal progenitor cells by several BMPs, ATDC5 embryonal carcinoma cells by GDF5 and BMP4-dependent lymphohematopoietic (CD34+CD31hi and CD34+CD31lo) progenitor cell development from ES cells. CHL2 may inhibits BMPs activity by blocking their interaction with their receptors, and has a negative regulator effect on the cartilage formation/regeneration from immature mesenchymal cells, by preventing or reducing the rate of matrix accumulation. Also, it may play a role during myoblast and osteoblast differentiation, and maturation.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.