Order Entry
Puerto Rico
ContactUsLinkComponent
 
Mouse Recombinant CD5
Catalog # 75791-682
Supplier:  Prosci
CAS Number:  
Mouse Recombinant CD5
Catalog # 75791-682
Supplier:  Prosci
Supplier Number:  92-560
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Species:
    Mouse
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage Conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein Synonyms:
    CD5 antigen (p56-62)|CD5|CD5 molecule|Lymphocyte antigen T1|CD5 antigen|Ly-1|LEU1T-cell surface glycoprotein CD5|T-cell surface glycoprotein CD5|Leu-1|T1
  • Protein/Peptide Name:
    CD5
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    38.9 kD
  • Sequence:
    Gln24-Asn370
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Cat. No.:
    75791-682
  • Supplier no.:
    92-560

Specifications

About this item

CD5 is a transmembrane glycoprotein of the conserved scavenger receptor cysteine-rich (SRCR) superfamily and expressed on thymocytes, peripheral T cells and a subset of B cells (B1-a). Moreover, CD5 also was found expressed in small lymphocytic lymphoma, hairy cell leukaemia and mantle cell lymphoma cells. The long cytoplasmic tail of CD5 has no intrinsic enzymatic activity, but contains four tyrosine phosphorylation sites, including an immunoreceptor tyrosine-based (ITAM)-like motif (pseudo-ITAM) and an immunoreceptor tyrosine-based inhibitory (ITIM)-like motif (pseudo-ITIM), as well as multiple potential serine and threonine phosphorylation sites. It physically associates with the T cell antigen receptor (TCR) and B cell antigen receptor (BCR), where it negatively modulates the activation and differentiation signals transduced by these receptors. CD5 also plays an important role in protection from activation-induced cell death and in the recognition of pathogen associated molecular patterns (PAMPS) present on fungal surfaces.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.