Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant Carboxylesterase 2E
Mouse Recombinant Carboxylesterase 2E
Catalog # 75791-642
Supplier:  Prosci
CAS Number:  
Mouse Recombinant Carboxylesterase 2E
Catalog # 75791-642
Supplier:  Prosci
Supplier Number:  92-540
CAS Number:  

Specifications

  • Enzyme type:
    Recombinant
  • Species:
    Mouse
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage Conditions:
    Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme Name:
    Carboxylesterase 2E
  • Enzyme Synonyms:
    Pyrethroid hydrolase Ces2e, carboxylesterase 2E, Ces5, CES2E
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    62.9 kD
  • Sequence:
    Met1-His556
  • Formulation:
    Lyophilized from a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Cat. No.:
    75791-642
  • Supplier no.:
    92-540

Specifications

About this item

Carboxylesterase 5 (CES5), also called cauxin or CES7, is a member of carboxylesterases family which plays an important role in the hydrolysis of ester and amide bonds

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

Carboxylesterase is a type of enzyme that capable of hydrolyzing a variety of carboxylic acid esters and it's widely distributed in cells especially in mammalian liver. CES5 is with broad substrate specificity ranging from small molecule esters to longchain fatty acid esters and thioesters. It has been previously reported CES5 was in high concentrations in the urine (cauxin) of adult male cats, and within a protein complex of mammalian male epididymal fluids. Roles for CES5 may include regulating urinary levels of male cat pheromones, catalyzing lipid transfer reactions within mammalian male reproductive fluids, and protecting neural tissue from drugs and xenobiotics.