- Protein/Peptide Type:Recombinant
- Species:Mouse
- Size:0.05 mg
- Tag sequence:C-Fc tag
- Storage Conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Endotoxin Content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein Synonyms:CD273|PD-L2|Pdcd1lg2|Programmed Cell Death 1 ligand 2|PD-1 ligand 2|B7-DC|PDCD1 ligand 2
- Protein/Peptide Name:PDL2
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular Weight:49.7 kD
- Sequence:Leu20-Arg219
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Cat. No.:75791-564
- Supplier no.:92-497
Programmed cell death 1 ligand 2 (PD-L2), also known as butyrophilin B7-DC or PDCD1 ligand 2, belongs to the member of B7 family which can regulate the activation and tolerance of T cells. PD-L2 is one ligand for Programmed cell death 1(PD-1), and the other is PD-L1. These two ligands shares 34% aa sequence identity. Mouse PD-L2 gene encodes a 273 amino acids (aa) protein with a putative 19 aa signal peptide, a 201 aa extracellular region , a 21 aa transmembrane domain and a 32 aa cytoplasmic region. The mouse PD-L2 gene is highly expressed in heart, placenta, pancreas, lung and liver while expressed weakly in spleen, lymph nodes and thymus. Besides, the expression of PD-L2 gene can be induced on dendritic cells grown from peripheral blood mononuclear cells under CSF2 and IL4/interleukin-4 treatment, and up-regulated by IFNG/IFN-gamma stimulation in monocytes. PD-L2 usually functions in a PDCD1-independent manner and is involved in regulating costimulatory signal which is essential for T-cell proliferation and IFNG production. Recent studies demonstrate that the expression of PD-L2 on the tumor cells promotes CD8 T cell–mediated rejection of tumor cells, at both the induction and effector phase of antitumor immunity. Moreover, PD-L2 binds to PD-1 cells and enhances T cell killing in a PD-1–independent mechanism.
- Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.