Order Entry
Puerto Rico
ContactUsLinkComponent
 
Mouse Recombinant PDCD1
Catalog # 75791-560
Supplier:  Prosci
CAS Number:  
Mouse Recombinant PDCD1
Catalog # 75791-560
Supplier:  Prosci
Supplier Number:  92-495
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Species:
    Mouse
  • Size:
    0.05 mg
  • Tag sequence:
    C-Fc tag
  • Storage Conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein Synonyms:
    PD-1|mPD-1|CD279|Pdcd1|Programmed Cell Death protein 1
  • Protein/Peptide Name:
    PDCD1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    43.3 kD
  • Sequence:
    Leu25-Gln167
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM Tris,150mM NaCl,pH8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Cat. No.:
    75791-560
  • Supplier no.:
    92-495

Specifications

About this item

Programmed Death-1 (PD-1), firstly cloned from mouse T cell hybridoma 2B4.11, is one member of CD28/CTLA-4 superfamily. PD-1 belongs to type I transmembrane protein and acts as an important immunosuppressive molecule. This family also include members of CD28, CTLA-4 and ICOS.The mouse Programmed Death-1 protein, encoded by PD-1 gene, comprises four parts including a putative 20 aa signal peptide, a 149 aa extracellular region, a 21 aa transmembrane domain and a 98 aa cytoplasmic region. The cytoplamsic tail of PD-1 contains two structural motifs, an immunoreceptor tyrosine-based inhibitory motif (ITIM) and an immunoreceptor tyrosine-based switch motif (ITSM) formed by two tyrosine residues which make the difference in PD-1 signal mediating. Mouse PD-1 is expressed in thymus and shares about 69% aa sequence identity with human PD-1. Recently, programmed death-1 (PD-1) with its ligands, programmed death ligand B7H1 (PD-L1) and B7DC (PD-L2), was found to regulate T-cell activation and tolerance, upon ligand binding, inhibiting T-cell effector functions in an antigen-specific manner. PD-1 gene knocked out mice would induce some autoimmune diseases, which suggests that PD-1 acts as a co-inhibitory molecule actively participating in maintaining peripheral tolerance. Thus, PD-1 may be a useful target for the immunologic therapy of carcinoma,infection,autoimmune diseases as well as organ transplantation.

  • Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.