Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant VEGF R1 (from Cells)
Human Recombinant VEGF R1 (from Cells)
Catalog # 75791-552
Supplier:  Prosci
CAS Number:  
Human Recombinant VEGF R1 (from Cells)
Catalog # 75791-552
Supplier:  Prosci
Supplier Number:  92-491
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-Fc tag
  • Storage Conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein Synonyms:
    FLT-1|Tyrosine-protein kinase receptor FLT|Tyrosine-protein kinase FRT|VEGFR-1|Vascular endothelial growth factor receptor 1|Vascular permeability factor receptor|Fms-like tyrosine kinase 1
  • Protein/Peptide Name:
    VEGF R1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    109 kD
  • Sequence:
    Ser27-Asn756
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Cat. No.:
    75791-552
  • Supplier no.:
    92-491

Specifications

About this item

Human Vascular endothelial growth factor receptor 1(VEGFR-1, FLT-1) is a member of the the class III subfamily of receptor tyrosine kinases (RTKs) and Tyr protein kinase family and CSF-1/PDGF receptor subfamily. VEGFR-1 is widely expressed in human tissues including normal lung, placenta, liver, kidney, heart and brain tissues. It is specifically expressed in most of the vascular endothelial cellsand peripheral blood monocytes. VEGFR-1 contains seven Ig-like C2-type domains and one protein kinase domain. VEGFR-1is an essential receptor tyrosine kinase and plays an important role in theregulation of VEGF family-mediated vasculogenesis, angiogenesis, and lymphangiogenesis. It is also mediators of neurotrophic activity and regulators of hematopoietic development. VEGFR-1 is a receptor for VEGF, VEGFB and PGF. It has a tyrosine-protein kinase activity. Tyrosine-protein kinase that acts as a cell-surface receptor for VEGFA, VEGFB and PGF.It may play an essential role as a negative regulator of embryonic angiogenesis by inhibiting excessive proliferation of endothelial cells and promote endothelial cell proliferation, survival and angiogenesis in adulthood. Its function in promoting cell proliferation seems to be cell-type specific. VEGFR-1 can also promote PGF-mediated proliferation of endothelial cells, proliferation of some types of cancer cells, but does not promote proliferation of normal fibroblasts (in vitro).

  • Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.