Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant Aldehyde dehydrogenase 3-A1 (from Cells)
Catalog # 75791-488
Supplier:  Prosci
CAS Number:  
Human Recombinant Aldehyde dehydrogenase 3-A1 (from Cells)
Catalog # 75791-488
Supplier:  Prosci
Supplier Number:  92-459
CAS Number:  

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage Conditions:
    Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme Name:
    Aldehyde dehydrogenase 3-A1
  • Enzyme Synonyms:
    Aldehyde dehydrogenase, dimeric NADP-preferring, ALDH3, ALDH3A1, Aldehyde dehydrogenase family 3 member A1, Aldehyde dehydrogenase 3, ALDHIII
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    51.4 kD
  • Sequence:
    Met1-His453, C-6 H
  • Formulation:
    Lyophilized from a 0.2 µm filtered solution of PBS,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Cat. No.:
    75791-488
  • Supplier no.:
    92-459

Specifications

About this item

Aldehyde dehydrogenase, dimeric NADP-preferring is an enzyme that in humans is encoded by the ALDH3A1 gene, belongs to the aldehyde dehydrogenase family

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

ALDHs play a major role in the detoxification of alcohol-derived acetaldehyde. They are involved in the metabolism of corticosteroids, biogenic amines, neurotransmitters, and lipid peroxidation. This protein preferentially oxidizes aromatic aldehyde substrates. It may play a role in the oxidation of toxic aldehydes.