Mouse Recombinant Carboxypeptidase A2
Catalog # 75791-408
Supplier: Prosci
CAS Number:
Specifications
- Enzyme type:Recombinant
- Species:Mouse
- Size:0.05 mg
- Tag sequence:C-6 His tag
- Storage Conditions:Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
- Endotoxin Content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Enzyme Name:Carboxypeptidase A2
- Enzyme Synonyms:CPA2/Carboxypeptidase A2, CPA2
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular Weight:46.2 kD
- Sequence:Gln17-Tyr417
- Formulation:Lyophilized from a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested Applications:Biological assays
- Cat. No.:75791-408
- Supplier no.:92-419
Specifications
About this item
Mouse carboxypeptidase A2(CPA2) is a secreted pancreatic procarboxy -peptidase which belongs to the peptidase M14 family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
CPA2 consists of a signal peptide, a pro region and a mature chain. It can be activated after cleavage of the pro peptide. CPA2 cleaves the C-terminal amide or ester bond of peptides that have a free C-terminal carboxyl group. The hydrolytic action of CPA2 was identified with a preference towards long substrates with aromatic amino acids in their C-terminal end, particularly tryptophan.