Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Human Recombinant CTGF (from Cells)

Human Recombinant CTGF (from Cells)

0.05 mg
Catalog # 75791-220
Supplier:   Prosci
Supplier #:  92-317
Human Recombinant CTGF (from Cells)
Catalog # 75791-220
Supplier:  Prosci
CAS Number:  
Human Recombinant CTGF (from Cells)

0.05 mg

CAS Number:  
Price
$958.78
Log in for availability
Each (50µG)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

CTGF belongs to the CCN (CTGF/Cyr61/Cef10/NOVH) protein family, which is comprised of six secreted proteins that reside in the extracellular matrix (ECM). CTGF causes a variety of cellular responses including reduced cell adhesion and enhanced cell migration and proliferation. CTGF has also been shown to be essential for epithelial to mesenchymal transition (EMT), a process whereby normal functioning cells morph into ones that produce mainly scar tissue (of which collagen in the major protein component).

  • Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.

Documents

Specifications

Specifications

  • Size:
    0.05 mg
  • Storage Conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Molecular Weight:
    62.6 kD
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Tested Applications:
    Bioassay
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Sequence:
    Gln27-Ala349
  • Source:
    Cells
  • Cat. No.:
    75791-220
  • Protein/Peptide Type:
    Recombinant
  • Gene ID:
    1490
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Protein Synonyms:
    IGFBP8|Hypertrophic chondrocyte-specific protein 24|Insulin-like growth factor-binding protein 8|CCN2|CCN family member 2|CTGF|Connective tissue growth factor
  • Protein/Peptide Name:
    CTGF
  • Tag sequence:
    C-Fc tag
  • Species:
    Human

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.