Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant S100 calcium binding A8 (from E. coli)
Mouse Recombinant S100 calcium binding A8 (from E. coli)
Catalog # 75791-160
Supplier:  Prosci
CAS Number:  
Mouse Recombinant S100 calcium binding A8 (from E. coli)
Catalog # 75791-160
Supplier:  Prosci
Supplier Number:  92-283
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage Conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein Synonyms:
    Mrp8|Chemotactic cytokine CP-10|Protein S100-A8|MRP-8|Leukocyte L1 complex light chain|S100 calcium-binding protein A8|Caga|S100a8|Calgranulin-A|Pro-inflammatory S100 cytokine|Migration inhibitory factor-related protein 8
  • Protein/Peptide Name:
    S100 calcium binding A8
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    11.3 kD
  • Sequence:
    Met1-Glu89
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mMTris,pH8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested Applications:
    Bioassay
  • Cat. No.:
    75791-160
  • Supplier no.:
    92-283

Specifications

About this item

Protein S100-A8(Mrp8) contains 2 EF-hand domains and belongs to the S-100 family. Mrp8 binds two calcium ions per molecule with an affinity similar to that of the S-100 proteins. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. It may function in the inhibition of casein kinase and as a cytokine. Altered expression of this protein is associated with the disease cystic fibrosis.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.