Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Storage Conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Endotoxin Content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein Synonyms:IL-TIF|IL-22|Cytokine Zcyto18|IL-10-related T-cell-derived-inducible factor|IL22|ILTIF|Interleukin-22|ZCYTO18
- Protein/Peptide Name:IL22
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular Weight:16.9 kD
- Sequence:Ala34-Ile179
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested Applications:Bioassay
- Cat. No.:75791-126
- Supplier no.:92-266
Specifications
About this item
Interleukin-22(IL-22) is a member of a group of the IL-10 family, a class of potent mediators of cellular inflammatory responses. IL-22 is produced by activated DC and T cells. IL-22 and IL-10 receptor chains play a role in cellular targeting and signal transduction. It can initiate and regulate innate immune responses against bacterial pathogens especially in epithelial cells such as respiratory and gut epithelial cells. IL-22 along with IL-17 likely plays a role in the coordinated response of both adaptive and innate immune systems. IL-22 also promotes hepatocyte survival in the liver and epithelial cells in the lung and gut similar to IL-10. Biological activity of IL-22 is initiated by binding to a cell-surface complex consisting of IL-22R1 and IL-10R2 receptor chains. IL-22 biological activity is further regulated by interactions with a soluble binding protein, IL-22BP. IL-22BP and an extracellular region of IL-22R1 share sequence similarity. In some cases, the pro-inflammatory versus tissue-protective functions of IL-22 are regulated by cytokine IL-17A.
This recombinant protein can be used for biological assays. For research use only.