Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant Carbonic Anhydrase 14 (from E. coli)
Catalog # 75790-892
Supplier:  Prosci
CAS Number:  
Human Recombinant Carbonic Anhydrase 14 (from E. coli)
Catalog # 75790-892
Supplier:  Prosci
Supplier Number:  92-149
CAS Number:  

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    N-6 His tag
  • Storage Conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme Name:
    Carbonic Anhydrase 14
  • Enzyme Synonyms:
    Carbonic Anhydrase 14, Carbonate Dehydratase XIV, Carbonic Anhydrase XIV, CA-XIV, CA14, UNQ690/PRO1335
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    32.8 kD
  • Sequence:
    Gly19-Met290
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM Tris, 150 mM NaCl, 10% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested Applications:
    Biological assays
  • Cat. No.:
    75790-892
  • Supplier no.:
    92-149

Specifications

About this item

Carbonic Anhydrase 14 (CA14) belongs to the Alpha-Carbonic Anhydrase family

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag

It is highly expressed in all parts of the central nervous system and lowly expressed in adult liver, heart, small intestine, colon, kidney, urinary bladder, and skeletal muscle. CA14 along with other Carbonic Anhydrases (CAs) participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. CA14 is predicted to be a type I membrane protein and catalyzes the reversible hydration of carbon dioxide.