Order Entry
United States
ContactUsLinkComponent
Human Recombinant Selenocysteine Lyase (from E. coli)
Human Recombinant Selenocysteine Lyase (from E. coli)
Catalog # 75790-630
Supplier:  Prosci
CAS Number:  
Human Recombinant Selenocysteine Lyase (from E. coli)
Catalog # 75790-630
Supplier:  Prosci
Supplier Number:  92-018
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Environmentally Preferable:
  • Tag sequence:
    N-6 His tag
  • Storage Conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme Name:
    Selenocysteine Lyase
  • Enzyme Synonyms:
    Selenocysteine Lyase, hSCL, SCLY, SCL
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    50.34 kD
  • Sequence:
    Met1-Ala445
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested Applications:
    Bioassay
  • Cat. No.:
    75790-630
  • Supplier no.:
    92-018

Specifications

About this item

Selenocysteine Lyase belongs to the class-V pyridoxal-phosphate-dependent aminotransferase family

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag

Selenocysteine Lyase exists as a homodimer in the cytosol. In the brain, Selenocysteine Lyase is as an enzyme that putatively salvages Sec and recycles the selenium for selenoprotein translation. Selenocysteine Lyase catalyzes the decomposition of L-selenocysteine to L-alanine and elemental selenium. Selenocysteine Lyase can be up-regulated In acute glomerulonephritis, it can also be regulated by JUN/AP-1.