Human Recombinant ULBP-2 (from Cells)
Catalog # 75790-466
Supplier: Prosci
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Source:Cells
- Species:Human
- Size:0.05 mg
- Tag sequence:C-Fc tag
- Storage Conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Endotoxin Content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein Synonyms:NKG2DL2|ULBP2|UL16-Binding Protein 2|N2DL2|NKG2D Ligand 2|N2DL-2|Retinoic Acid Early Transcript 1H|ALCAN-Alpha|RAET1H
- Protein/Peptide Name:ULBP-2
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular Weight:51.4 kD
- Sequence:Gly26-Ser217
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested Applications:used for biological assays
- Cat. No.:75790-466
- Supplier no.:91-915
Specifications
About this item
NKG2D Ligand 2 (N2DL2) is a member of a family of cell-surface proteins. N2DL2 function as ligands for human cytomegalovirus glycoprotein UL16. N2DL2 is anchored to the membrane via a GPI-linkage. N2DL2 is bind to human NKG2D, an activating receptor expressed on NK cells, NKT cells, T cells. Engagement of NKG2D results in the activation of cytolytic activity and cytokine production by these effects cells. The ULBPs are expressed on some tumor cells and have been implicated in tumor surveillance.
- Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.