Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Carboxypeptidase E (from Cells)
Human Recombinant Carboxypeptidase E (from Cells)
Catalog # 75790-332
Supplier:  Prosci
CAS Number:  
Human Recombinant Carboxypeptidase E (from Cells)
Catalog # 75790-332
Supplier:  Prosci
Supplier Number:  91-840
CAS Number:  

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage Conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme Name:
    Carboxypeptidase E
  • Enzyme Synonyms:
    Carboxypeptidase E(CPE for short), Carboxypeptidase H, Enkephalin convertase, Prohormone-processing carboxypeptidase, CPE
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    47.1 kD
  • Sequence:
    Arg42-Ser453
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl,1 mM ZnCl₂, 10% Glycerol,pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested Applications:
    Bioassay
  • Cat. No.:
    75790-332
  • Supplier no.:
    91-840

Specifications

About this item

The active form of CPE cleaves C-terminal amino acid residues of the peptide, and is thus involved in the biosynthesis of peptide hormones and neurotransmitters including insulin, enkephalin, etc

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

It is thought that membrane-associated CPE acts as a sorting receptor for targeting regulated secretory proteins which are mostly prohormones and neuropeptides in the trans-Golgi network of the pituitary and in secretory granules into the secretory pathway. Defects in this protein are implicated in type II diabetes due to impaired glucose clearance and insulin resistance.