Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant Collectin 11 (from Cells)
Catalog # 75790-174
Supplier:  Prosci
CAS Number:  
Human Recombinant Collectin 11 (from Cells)
Catalog # 75790-174
Supplier:  Prosci
Supplier Number:  91-761
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage Conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein Synonyms:
    Collectin Kidney Protein 1|COLEC11|Collectin-11|CL-K1
  • Protein/Peptide Name:
    Collectin 11
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    27.14 kD
  • Sequence:
    Gln26-Met271
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested Applications:
    Bioassay
  • Cat. No.:
    75790-174
  • Supplier no.:
    91-761

Specifications

About this item

Collectin-11 is a secreted protein that belongs to the COLEC10/COLEC11 family. Collectin-11 contains one C-type lectin domain and one collagen-like domain. Collectins play important roles in the innate immune system by binding to carbohydrate antigens on microorganisms, facilitating their recognition and removal. Collectin-11 binds to various sugars including fucose and mannose, but does not bind to glucose, N-acetylglucosamine and N-acetylgalactosamine. It has a higher affinity for fucose compared to mannose. Collectin-11 binds lipopolysaccharides (LPS). It also involved in fundamental development serving as a guidance cue for neural crest cell migration. Defects in Collectin-11 are the cause of 3MC syndrome type 2 (3MC2).

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.