Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Calcitonin (from Cells)
Human Recombinant Calcitonin (from Cells)
Catalog # 75790-122
Supplier:  Prosci
CAS Number:  
Human Recombinant Calcitonin (from Cells)
Catalog # 75790-122
Supplier:  Prosci
Supplier Number:  91-731
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage Conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein Synonyms:
    PDN-21|CCP|CALC1|Calcitonin|Katacalcin|Calcitonin Carboxyl-Terminal Peptide|CALCA
  • Protein/Peptide Name:
    Calcitonin
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    13.8 kD
  • Sequence:
    Ala26-Asn141
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM TrisHCl, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested Applications:
    Bioassay
  • Cat. No.:
    75790-122
  • Supplier no.:
    91-731

Specifications

About this item

Calcitonin is a secreted protein which belongs to the calcitonin family. Calcitonin is cleaved into the following two chains: Calcitonin and Katacalcin. Katacalcin is a potent plasma calcium-lowering peptide. Calcitonin is a 32-amino acid linear polypeptide hormone. Calcitonin acts to reduce blood calcium (Ca2+), opposing the effects of parathyroid hormone (PTH). Its importance in humans has not been as well established as its importance in other animals, as its function is usually not significant in the regulation of normal calcium homeostasis. Calcitonin causes a rapid but short-lived drop in the level of calcium and phosphate in blood by promoting the incorporation of those ions in the bones.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.