Order Entry
United States
ContactUsLinkComponent
Human Recombinant GMP Reductase 1 (from Cells)
Human Recombinant GMP Reductase 1 (from Cells)
Catalog # 75789-992
Supplier:  Prosci
CAS Number:  
Human Recombinant GMP Reductase 1 (from Cells)
Catalog # 75789-992
Supplier:  Prosci
Supplier Number:  91-666
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Environmentally Preferable:
  • Tag sequence:
    C-6 His tag
  • Storage Conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme Name:
    GMP Reductase 1
  • Enzyme Synonyms:
    GMP Reductase 1, Guanosine 5'-Monophosphate Oxidoreductase 1, Guanosine Monophosphate Reductase 1, GMPR, GMPR1
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    38.5 kD
  • Sequence:
    Met1-Ser345
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM Tris-HCl, 40% glycerol, 0.15 M NaCl and 1 mM DTT, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH₂O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested Applications:
    Bioassay
  • Cat. No.:
    75789-992
  • Supplier no.:
    91-666

Specifications

About this item

GMP Reductase 1 (GMPR) is a member of the IMPDH/GMPR family

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

GMPR exists as a homotetramer and catalyzes the irreversible NADPH-dependent deamination of GMP to IMP. It functions in the conversion of nucleobase, nucleoside and nucleotide derivatives of G to A nucleotides, and in maintaining the intracellular balance of A and G nucleotides. GMP reductase gene expression may be regulated by MITF. At least two different alleles are known.