Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant Carboxypeptidase M
Mouse Recombinant Carboxypeptidase M
Catalog # 75789-904
Supplier:  Prosci
CAS Number:  
Mouse Recombinant Carboxypeptidase M
Catalog # 75789-904
Supplier:  Prosci
Supplier Number:  91-622
CAS Number:  

Specifications

  • Enzyme type:
    Recombinant
  • Species:
    Mouse
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage Conditions:
    Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme Name:
    Carboxypeptidase M
  • Enzyme Synonyms:
    Carboxypeptidase M, CPM
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    47.5 kD
  • Sequence:
    Leu18-Ser423
  • Formulation:
    Lyophilized from a 0.2 µm filtered solution of PBS, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested Applications:
    Bioassay
  • Cat. No.:
    75789-904
  • Supplier no.:
    91-622

Specifications

About this item

Carboxypeptidase M (CPM) belongs to the peptidase M14 family, and exists in cell membrane

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

The protein binds 1 zinc ion per subunit, and cleavage of C-terminal arginine or lysine residues from polypeptides. CPM specifically removes C-terminal basic residues (Arg or Lys) from peptides and proteins. It is believed to play important roles in the control of peptide hormone and growth factor activity at the cell surface, and in the membrane-localized degradation of extracellular proteins.