Human Recombinant CD300A (from Cells)
75789-704
: Prosci
:
- Protein/Peptide Type:Recombinant
- Source:Cells
- Species:Human
- Size:0.05 mg
- Tag sequence:C-6 His tag
- Storage Conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Endotoxin Content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein Synonyms:CMRF35H|CD300a|Immunoglobulin Superfamily Member 12|CMRF35-H9|CD300 antigen-like Family Member A|CLM-8|LMIR1|IRC1/IRC2|CMRF35-H|IgSF12|IRp60|CMRF-35-H9|Inhibitory Receptor Protein 60|NK Inhibitory Receptor|CMRF35-like Molecule 8
- Protein/Peptide Name:CD300A
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular Weight:18.48 kD
- Sequence:Leu18-Gln178
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested Applications:Bioassay
- Cat. No.:75789-704
- Supplier no.:91-514
CD300A is a single-pass type I membrane protein that belongs to the CD300 family. CD300A consists of a 163 amino acid (aa) extracellular domain (ECD) with one Ig-like V- type domain, a 21 amino acid transmembrane segment, and a 98 amino acid cytoplasmic domain with tyrosine residues. CD300A is expressed not only by natural killer (NK) cells but also by T-cell subsets, B-cells, dendritic cells, mast cells, granulocytes and monocytes. CD300A is an inhibitory receptor which may contribute to the down-regulation of cytolytic activity in natural killer (NK) cells, and to the down-regulation of mast cell degranulation.
- Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.