Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Vanin-1 (from Cells)
Human Recombinant Vanin-1 (from Cells)
Catalog # 75789-670
Supplier:  Prosci
CAS Number:  
Human Recombinant Vanin-1 (from Cells)
Catalog # 75789-670
Supplier:  Prosci
Supplier Number:  91-497
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage Conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein Synonyms:
    Vascular Non-Inflammatory Molecule 1|VNN1|Vanin-1|Tiff66|Pantetheinase|Pantetheine Hydrolase
  • Protein/Peptide Name:
    Vanin-1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    53.27 kD
  • Sequence:
    Gln22-Ser490
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested Applications:
    Bioassay
  • Cat. No.:
    75789-670
  • Supplier no.:
    91-497

Specifications

About this item

Vanin-1 is a cell membrane protein which contains one CN hydrolase domain and belongs to the CN hydrolase family and BTD/VNN subfamily. Vanin-1 is also a member of the Vanin family of proteins, which share extensive sequence similarity with each other, and also with biotinidase. The family includes secreted and membrane-associated proteins, a few of which have been reported to participate in hematopoietic cell trafficking. Vanin-1 is widely expressed with higher expression in spleen, kidney and blood and overexpressed in lesional psoriatic skin. No biotinidase activity has been demonstrated for any of the vanin proteins; however, they possess pantetheinase activity, which may play a role in oxidative-stress response. Vanin-1 is an epithelial pantetheinase that provides cysteamine to tissue and regulates response to stress. Vanin-1 is expressed by enterocytes, and its absence limits intestinal epithelial cell production of proinflammatory signals. Vanin-1 regulates late adhesion steps of thymus homing under physiological, noninflammatory conditions. The early impact of vanin-1 deficiency on tumor induction was directly correlated to the amount of inflammation and subsequent epithelial proliferation rather than cell death rate. Vanin-1 molecule was shown to be involved in the control of thymus reconstitution following sub-lethal irradiation.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.