Human Recombinant Aminopeptidase P2 (from Cells)
Catalog # 75789-596
Supplier: Prosci
CAS Number:
Specifications
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Size:0.05 mg
- Tag sequence:C-6 His tag
- Storage Conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Endotoxin Content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Enzyme Name:Aminopeptidase P2
- Enzyme Synonyms:Xaa-Pro Aminopeptidase 2, Aminoacylproline Aminopeptidase, Membrane-Bound Aminopeptidase P, Membrane-Bound APP, Membrane-Bound AmP, mAmP, X-Pro Aminopeptidase 2, XPNPEP2
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular Weight:71.64 kD
- Sequence:His22-Ala650
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested Applications:Bioassay
- Cat. No.:75789-596
- Supplier no.:91-460
Specifications
About this item
Xaa-Pro aminopeptidase 2 (XPNPEP2) belongs to the peptidase M24B family of metalloproteases
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Human XPNPEP2 is widely expressed in various tissues, such as kidney, lung, heart, placenta, liver, small intestine and colon. However, it doesn’t express in brain, skeletal muscle, pancreas, spleen, thymus, prostate, testis and ovary. XPNPEP2 is a Homotrimer which binds 2 manganese ions per subunit. The metalloprotease XPNPEP2 may play a role in the inflammatory process and other reactions produced in response to injury or infection and the metabolism of the vasodilator bradykinin.