Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant OSM R beta (from Cells)
Human Recombinant OSM R beta (from Cells)
Catalog # 75789-572
Supplier:  Prosci
CAS Number:  
Human Recombinant OSM R beta (from Cells)
Catalog # 75789-572
Supplier:  Prosci
Supplier Number:  91-448
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage Conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein Synonyms:
    OSMR|Interleukin-31 Receptor Subunit Beta|IL-31 Receptor Subunit Beta|IL-31R Subunit Beta|OSMRB|Oncostatin-M-Specific Receptor Subunit Beta|IL-31RB|IL-31R-Beta
  • Protein/Peptide Name:
    OSM R beta
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    82.03 kD
  • Sequence:
    Glu28-Ser739
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested Applications:
    For Biological Assays
  • Cat. No.:
    75789-572
  • Supplier no.:
    91-448

Specifications

About this item

Oncostatin-M-Specific Receptor Subunit beta (OSMR beta ) is a 150 - 180 kDa member of the IL-6 receptor family. OSMR beta associates with gp130 to form the type II OSM receptor, the receptor is responsive to OSM. Gp130 subunit is shared by other IL-6 family cytokine receptors, and OSMR beta associates with gp130-like receptor (GPL) to form a receptor complex responsive to IL-31. The human OSMR beta cDNA encodes a 979 amino acid (aa) precursor, the precursor includes a 27 aa signal sequence, a 712 aa extracellular domain (ECD), a 22 aatransmembrane segment, and a 218 aa cytoplasmic domain. The ECD contains one partial and one complete hematopoietin domain, an Ig-like domain, and three Fibronectin type-III domains.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.