Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Carboxylesterase 1 (from Cells)
Human Recombinant Carboxylesterase 1 (from Cells)
Catalog # 75789-488
Supplier:  Prosci
CAS Number:  
Human Recombinant Carboxylesterase 1 (from Cells)
Catalog # 75789-488
Supplier:  Prosci
Supplier Number:  91-406
CAS Number:  

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage Conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme Name:
    Carboxylesterase 1
  • Enzyme Synonyms:
    Liver Carboxylesterase 1, Acyl-Coenzyme A:Cholesterol Acyltransferase, ACAT, Brain Carboxylesterase hBr1, Carboxylesterase 1, CE-1, hCE-1, Cocaine Carboxylesterase, Egasyn, HMSE, Methylumbelliferyl-Acetate Deacetylase 1, Monocyte/Macrophage Serine Esterase, Retinyl Ester Hydrolase, REH, Serine Esterase 1, Triacylglycerol Hydrolase, TGH, CES1, CES2, SES1
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    61.05 kD
  • Sequence:
    His19-Glu562
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM HAc-NaAc, 150 mM NaCl, pH 4.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested Applications:
    Bioassay
  • Cat. No.:
    75789-488
  • Supplier no.:
    91-406

Specifications

About this item

Carboxylesterase 1 (CES1) is a member of a large family of carboxylesterases that are responsible for the hydrolysis of ester and amide bonds

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

These enzymes have broad substrate specificity ranging from small molecule esters such as phenylester to long chain fatty acid esters and thioesters. They are major determinants of the pharmacokinetic behavior of most therapeutic agents containing an ester or amide bond. CES1 shares the serine hydrolase fold observed in other esterases. CES1 hydrolyzes aromatic and aliphatic esters, but has no catalytic activity toward amides or a fatty acyl-CoA ester. CES1 participates in detoxification of drugs such as cocaine and heroin in serum and liver. It may also play a role in detoxification in the lung and/or protection of the central nervous system from ester or amide compounds.