Human Recombinant LMIR2 (from Cells)
Catalog # 75789-474
Supplier: Prosci
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Source:Cells
- Species:Human
- Size:0.05 mg
- Tag sequence:C-6 His tag
- Storage Conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Endotoxin Content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein Synonyms:CMRF35|Immunoglobulin Superfamily Member 16|CLM-6|CMRF35-A1|CD300 Antigen-like Family Member C|IgSF16|CMRF-35|CMRF35-like Molecule 6|CD300c|CMRF35A|CMRF35A1
- Protein/Peptide Name:LMIR2
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular Weight:18.89 kD
- Sequence:Gly21-Arg183
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested Applications:Bioassay
- Cat. No.:75789-474
- Supplier no.:91-399
Specifications
About this item
CD300C is a single-pass type I membrane protein which belongs to the immunoregulatory signaling (IRS) family. CD300C contains one Ig-like V-type domain and is present on the surface of natural killer cells, granulocytes, most myeloid cells, dendritic cells, and a subpopulation of T and B lymphocytes. The CD300C (CMRF-35A) and CD300A (CMRF-35H) molecules are homologous leukocyte surface proteins. CD300a and CD300C play an important role in the cross-regulation of TNF-alpha and IFN-alpha secretion from pDCs. CD300A and CD300C are indistinguishable on the surface of NK cells. The ligand for CD300C is presently unknown.
- Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.