- Protein/Peptide Type:Recombinant
- Source:Cells
- Species:Human
- Size:0.05 mg
- Tag sequence:C-6 His tag
- Storage Conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Endotoxin Content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein Synonyms:ON|Basement-Membrane Protein 40|SPARC|Osteonectin|Secreted Protein Acidic and Rich in Cysteine|BM-40
- Protein/Peptide Name:SPARC
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular Weight:33.73 kD
- Sequence:Ala18-Ile303
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested Applications:For Biological Assays
- Cat. No.:75789-386
- Supplier no.:91-355
Secreted Protein Acidic and Rich in Cysteine (SPARC) is a secreted, evolutionarily conserved collagen-binding glycoprotein and belongs to the SPARC family. SPARC has 286 amino acids and contains an EF-hand in C-termina domain, a follistatin-like domain with Kazal-like sequences. There are two calcium binding sites, one binds 5 - 8 Ca2+ with a low affinity and other on an EF-hand loop that binds a Ca2+ ion with a high affinity. It is highly expressed in tissues undergoing morphogenesis, remodeling and wound repair. SPARC regulate cell growth through interactions with the extracellular matrix (ECM) and cytokines. SPARC bind to numerous proteins of the ECM, affect ECM protein expression, influence cellular adhesion and migration, and modulate growth factor-induced cell proliferation and angiogenesis. SPARC also binds several types of collagen, albumin, thrombospondin, PDGF and cell membranes.
- Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.