Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant UGRP1 (from E. coli)
Human Recombinant UGRP1 (from E. coli)
Catalog # 75789-056
Supplier:  Prosci
CAS Number:  
Human Recombinant UGRP1 (from E. coli)
Catalog # 75789-056
Supplier:  Prosci
Supplier Number:  91-170
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Storage Conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein Synonyms:
    UGRP1|Uteroglobin-Related Protein 1|PNSP1|Pneumo Secretory Protein 1|SCGB3A2|PnSP-1|Secretoglobin Family 3A Member 2
  • Protein/Peptide Name:
    UGRP1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    7.9 kD
  • Sequence:
    Phe22-Val93
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested Applications:
    Bioassay
  • Cat. No.:
    75789-056
  • Supplier no.:
    91-170

Specifications

About this item

Uteroglobin-Related Protein 1 (UGRP1) belongs to the secretoglobin family which has been suggested to play a role in lung inflammation and allergic diseases. UGRP1 is a 17 kDa secreted homodimeric protein that shows amino acid sequence similarity with uteroglobin. UGRP1 is expressed predominantly in the lung and low levels of expression are detected in the thyroid. Expression of UGRP1 in lung epithelial cells is enhanced by IL-10 and decreased through the activities of IL-9 and IL-5. UGRP1 interacts with the macrophage scavenger receptor with collagenous structure which is expressed by alveolar macrophages in the lung. It have suggested that UGRP1 may be involved in inflammation and pathogen clearance in the lung by binding to its receptor.

This recombinant protein can be used for biological assays. For research use only.