Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Stratifin (from E. coli)
Human Recombinant Stratifin (from E. coli)
Catalog # 75789-022
Supplier:  Prosci
CAS Number:  
Human Recombinant Stratifin (from E. coli)
Catalog # 75789-022
Supplier:  Prosci
Supplier Number:  91-153
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Storage Conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein Synonyms:
    14-3-3 Protein Sigma|Stratifin|SFN|Epithelial Cell Marker Protein 1|HME1
  • Protein/Peptide Name:
    Stratifin
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    27.77 kD
  • Sequence:
    Met1-Ser248
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 250mM NaCl, 1mM EDTA, 1mM DTT, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested Applications:
    For Biological assays
  • Cat. No.:
    75789-022
  • Supplier no.:
    91-153

Specifications

About this item

Stratifin (SFN) belongs to the 14-3-3 family of proteins that act as important regulators of intracelluar signal transduction through their ability to bind specific motifs phosphorylated on serine or threonine. There are at least seven isoforms that have been identified in mammals (beta, gamma, epsilon, sigma, zeta, tau and eta). SFN can detected in many tissues, highly expressed in stratified squamous keratinizing epithelium. SFN is indicated as an epithelial cell marker and serves as a tumor suppressor whose expression can be down regulated by methylation. In addition, SFN plays a key role in maintaining the G2 checkpoint in cells and preventing mitotic death.

This recombinant protein can be used for biological assays. For research use only.