Human Recombinant Retinol Binding 4 (from E. coli)
Catalog # 75789-014
Supplier: Prosci
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Storage Conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Endotoxin Content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein Synonyms:RBP4|RBP|Plasma Retinol-Binding Protein|Retinol-Binding Protein 4|PRBP
- Protein/Peptide Name:Retinol Binding 4
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular Weight:21.2 kD
- Sequence:Glu19-Leu201
- Formulation:Lyophilized from a 0.2 um filtered solution of 50mM TrisHCl, 10mM CaCl2, 150mM NaCl, pH 7.5 . Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested Applications:Bioassay
- Cat. No.:75789-014
- Supplier no.:91-149
Specifications
About this item
Retinol Binding Protein 4 (RBP4) is a member of the Lipocalin family and in the blood. RBP4 is the specific vector for retinol. RBP4 is expressed and secreted by adipose tissue, and is associated with insulin resistance. RBP4 delivers retinol from the liver stores to the peripheral tissues. In plasma, the RBP-retinol complex interacts with transthyretin to prevents its loss by filtration through the kidney glomeruli. Defects in RBP4 cause retinol-binding protein deficiency and can cause night vision problems.
This recombinant protein can be used for biological assays. For research use only.