Human Recombinant PTP1C (from E. coli)
75789-012
: Prosci
:
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Tag sequence:C-6 His tag
- Storage Conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Endotoxin Content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein Synonyms:SH-PTP1|Tyrosine-Protein Phosphatase Non-Receptor Type 6|PTP1C|Protein-Tyrosine Phosphatase 1C|Protein-Tyrosine Phosphatase SHP-1|PTP-1C|HCP|PTPN6|Hematopoietic Cell Protein-Tyrosine Phosphatase
- Protein/Peptide Name:PTP1C
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular Weight:35.38 kD
- Sequence:Lys243-Ile541
- Formulation:Supplied as a 0.2 um filtered solution of 25mM TrisHCl, 2mM β-ME, 1mM EDTA, 1mM DTT, 20% Glycerol, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested Applications:Bioassay
- Cat. No.:75789-012
- Supplier no.:91-148
Protein-Tyrosine Phosphatase 1C (PTP1C) belongs to the protein-tyrosine phosphatase family.which is known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. PTP1C is highly expressed in leukocyte cell type. It contains two SH2 domains and one tyrosine-protein phosphatase domain. The SH2 regions may interact with other cellular components to modulate its own phosphatase activity against interacting substrates. In addition, PTP1C also modulates signaling by tyrosine phosphorylated cell surface receptors.
- Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.