Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Human Recombinant Esterase D (from <i>E. coli</i>)

Human Recombinant Esterase D (from E. coli)

0.05 mg
Catalog # 75788-946
Supplier:   Prosci
Supplier #:  91-115
Human Recombinant Esterase D (from <i>E. coli</i>)
Catalog # 75788-946
Supplier:  Prosci
CAS Number:  
Human Recombinant Esterase D (from E. coli)

0.05 mg

CAS Number:  
Price
$743.14
Log in for availability
Each (50µG)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Human Esterase D is a serine hydrolase that is involved in the detoxification of formaldehyde

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

Esterase D plays a part in a variety of substrates, including O-acetylated sialic acids, which may involves in the recycling of sialic acids. Esterase D can be used as a genetic marker for retinoblastoma and Wilson’s disease.

Documents

Specifications

Specifications

  • Size:
    0.05 mg
  • Storage Conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme Synonyms:
    S-Formylglutathione Hydrolase, FGH, Esterase D, Methylumbelliferyl-Acetate Deacetylase, ESD
  • Enzyme type:
    Recombinant
  • Molecular Weight:
    32.59 kD
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Tested Applications:
    Bioassay
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Sequence:
    Met1-Ala282
  • Enzyme Name:
    Esterase D
  • Source:
    E. coli
  • Cat. No.:
    75788-946
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 10% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tag sequence:
    C-6 His tag
  • Species:
    Human

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.