Specifications
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Tag sequence:C-6 His tag
- Storage Conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Endotoxin Content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Enzyme Name:Carbonic Anhydrase 1
- Enzyme Synonyms:Carbonic Anhydrase 1, Carbonate Dehydratase I, Carbonic Anhydrase B, CAB, Carbonic Anhydrase I, CA-I, CA1
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular Weight:29.93 kD
- Sequence:Ala2-Phe261
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, 10% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested Applications:For Biological assays
- Cat. No.:75788-904
- Supplier no.:91-090
Specifications
About this item
Carbonic Anhydrase 1 (CA1) is a cytosolic enzyme, belonging to the alpha-carbonic anhydrase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
It is highly expressed in erythrocytes and acts as an early marker for erythroid differentiation. Carbonic anhydrase 1 plays a improtant role in many biological processes such as calcification, cellular respiration, bone resorption, acid-base balance. It is activated by imidazole, histamine, L-adrenaline, L- and D-histidine, and L- and D-phenylalanine. At the same time, It is inhibited by sulfonamide derivatives and coumarins. In addition, CA1 is a zinc metalloenzyme that has reversible hydration of carbon dioxide. It can hydrate cyanamide to urea.