Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant IL22 (from E. coli)
Mouse Recombinant IL22 (from E. coli)
Catalog # 75788-798
Supplier:  Prosci
CAS Number:  
Mouse Recombinant IL22 (from E. coli)
Catalog # 75788-798
Supplier:  Prosci
Supplier Number:  91-037
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    0.05 mg
  • Storage Conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein Synonyms:
    IL-TIF Alpha|IL-TIF|IL-22|IL-22a|IL-10-Related T-Cell-Derived-Inducible Factor|Il22a|Iltif|Interleukin-22a|Interleukin-22|Il22|Iltifa
  • Protein/Peptide Name:
    IL22
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    16.7 kD
  • Sequence:
    Leu34-Val179
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested Applications:
    For Biological assays
  • Cat. No.:
    75788-798
  • Supplier no.:
    91-037

Specifications

About this item

Interleukin-22 (IL-22) was initially identified as a gene induced by IL-9 in mouse T cells and mast cells. Mouse IL-22 cDNA encodes a 179 amino acid residue protein with a putative 33 amino acid signal peptide that is cleaved to generate a 147 amino acid mature protein that shares approximately 79% and 22% sequence identity with human IL22 and IL10, respectively. IL22 has been shown to activate STAT-1 and STAT-3 in several hepatoma cell lines and up-regulate the production of acute phase proteins. IL-22 is produced by normal mouse T cells upon Con A activation. Mouse IL-22 expression is also induced in various organs upon lipopolysaccharide injection, suggesting that IL-22 may be involved in inflammatory responses. The functional IL-22 receptor complex consists of two receptor subunits, IL-22R (previously an orphan receptor named CRF2-9) and IL-10R beta (previously known as CRF2-4), belonging to the class II cytokine receptor family.

This recombinant protein can be used for biological assays. For research use only.