Orders LinkContactUsLinkComponent
Ward's® Mineral Thin Sections Set
Ward's® Mineral Thin Sections Set
Mineral Thin Sections Set, 40
Catalog # 470024-866
Supplier:   Avantor
Ward's® Mineral Thin Sections Set
Catalog # 470024-866
Supplier:  Avantor
Ward's® Mineral Thin Sections Set
Ward's® Mineral Thin Sections Set
Mineral Thin Sections Set, 40
Price
$2,499.99
Log in for availability
Each KIT(1items) Kit of 1
Product Details & Documents
Forty mineral thin sections for Petrography Studies
  • Pure Mineral samples
  • Hand Made by Wards® staff Geologists
Our excellent series of forty standard thin sections features important and interesting rock-forming mineral types most often found in field samples.
Each thin section is made from pure mineral samples or from rocks containing a good amount of the mineral. This set is housed in standard slide boxes and an identification list is included.
Specifications
  • Description:
    Mineral Thin Sections Set
  • No. of slides:
    40