Lab Freezers, General Purpose Series, Thermo Scientific
Supplier: Avantor
Certificates
About this item
Store samples from general purpose applications in these general-purpose lab freezers, which feature reliable cold storage and are highly dependable.
- Digital temperature controller with intuitive interface
- Excellent temperature stability of ±3°C
- High-density, fluorine-free insulation
- Automatic defrost
- Optional chart recorder
- High/Low temperature, visual/audio alarms
- Easily accessible contacts for backup monitoring
Temperature range: -12 to -30 °C
Certifications: UL listed.
Frequently Bought Together