Order Entry
Puerto Rico
ContactUsLinkComponent
Lactate Dehydrogenase (Hitachi & Beckman) Reagent Set, Pointe Scientific
Lactate Dehydrogenase (Hitachi & Beckman) Reagent Set, Pointe Scientific
Catalog # 10022-958
CAS Number:  
Lactate Dehydrogenase (Hitachi & Beckman) Reagent Set, Pointe Scientific
Catalog # 10022-958
Supplier Number:  HL956-295
CAS Number:  

Specifications

  • Description:
    Hitachi Lactate Dehydrogenase
  • Size:
    210 mL R1 Reagent
    85 mL R2 Reagent
  • Cat. No.:
    10022-958
  • Format:
    Liquid
  • Storage temperature:
    2...8°C (35.6...46.4 °F)
  • Supplier no.:
    HL956-295

Specifications

About this item

The Lactate Dehydrogenase (Hitachi & Beckman) Reagent Set is for the quantitative kinetic determination of lactate dehydrogenase activity in serum.

  • Quantitative kinetic determination of lactate dehydrogenase activity in serum
  • Catalyzes the oxidation of lactate to pyruvate with simultaneous reduction of NAD to NADH
  • For in vitro diagnostic use only

The Lactate Dehydrogenase (Hitachi & Beckman) Reagent Set is for the quantitative kinetic determination of lactate dehydrogenase activity in serum

The Hitachi 295 mL reagent is for use on the Hitachi 917 analyzer only and for in vitro diagnostic use only. There are five 42 mL R1 Reagent bottles and five 17 mL R2 Reagent bottles in this kit.

The Hitachi 568 mL reagent is for use on Hitachi 717, 911, 912, and 914 analyzers and for in vitro diagnostic use only. There are four 100 mL R1 Reagent bottles and eight 21 mL R2 Reagent bottles in this kit.

The Olympus reagent is for use on the Beckman Coulter AU 400, 480, and 640 analyzers and is for in vitro diagnostic use only. There are five 42 mL R1 Reagent bottles and five 17 mL R2 Reagent bottles in the Olympus kit.

Certifications: CE Certified
Ordering information: Materials Required but not Provided:
Control sera with known normal and abnormal Lactate Dehydrogenase values.