About this item
The Total Bilirubin (Hitachi & Beckman) Reagent Set is for the quantitative determination of total bilirubin in serum.
- Quantitative determination of total bilirubin in serum
- In vitro diagnostic use only
- The intensity of the color of the azobilirubin produced can be measured photometrically.
The Total Bilirubin (Hitachi & Beckman) Reagent Set is for the quantitative determination of total bilirubin in serum
The Hitachi 711 mL reagent is for use on the Hitachi 917 analyzer only and for in vitro diagnostic use only. There are nine 63 mL R1 Reagent bottles and nine 16 mL R2 Reagent bottles in this kit.
The Hitachi 639 mL reagent is for use on Hitachi 717, 911, 912, and 914 analyzers and for in vitro diagnostic use only. There are six 90 mL R1 Reagent bottles and three 51 mL R2 Reagent bottles in this kit.
The Olympus reagent is for use on the Beckman Coulter AU 400, 480, and 640 analyzers and is for in vitro diagnostic use only. There are nine 63 mL R1 Reagent bottles and nine 16 mL R2 Reagent bottles in this kit.
Control sera with known normal and abnormal direct bilirubin values as well as Calibrator.