Tris Borate EDTA Buffer, for Molecular Biology, 10X, DNAse, RNAse and Protease free, pH 8.3, Thermo Scientific
Catalog # TS32733-0010
Supplier: Thermo Scientific Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Plastic bottle
- Description:TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer)
- Size:1 L
- Cat. No.:TS32733-0010
Specifications
About this item
CAS No.: 610769-35-2
- MDL No.: MFCD00004679
- GHS Symbols: GHS07
- Appearance: Clear colorless Solution
- Storage Temperature: Ambient
- Signal word: Warning