BUN (Urea Nitrogen) Reagent Set, Pointe Scientific
Supplier: HORIBA INSTRUMENTS INCORPORATED
Certificates
About this item
BUN (Urea Nitrogen) Reagent Set is used for the quantitative determination of urea nitrogen in serum.
- Quantitative determination of urea nitrogen in serum
- in vitro diagnostic use only
- Liberated ammonia reacts with α-ketoglutarate in the presence of NADH to yield glutamate
BUN (Urea Nitrogen) Reagent Set is used for the quantitative determination of urea nitrogen in serum
Hitachi BUN 654 ml reagent is for use on the Hitachi 917 analyzer only and for in vitro diagnostic use only. There are six 66 mL R1 Reagent bottles and six 43 mL R2 Reagent bottles in this kit.
Hitachi BUN 1L reagent is for use on Hitachi 717, 911, 912, and 914 analyzers and for in vitro diagnostic use only. There are eight 100 mL R1 Reagent bottles and four 50 mL R2 Reagent bottles in this kit.
Certifications: CE Certified
Ordering information: Materials Required but not Provided:
An NIST-traceable BUN standard (B7550-STD) or serum calibrator (C7506-50) as well as a control sera with known normal and abnormal BUN values (C7590/91/92).
An NIST-traceable BUN standard (B7550-STD) or serum calibrator (C7506-50) as well as a control sera with known normal and abnormal BUN values (C7590/91/92).