ALT Liquid Reagent Set, Pointe Scientific
Supplier: HORIBA INSTRUMENTS INCORPORATED
Certificates
About this item
ALT Liquid Reagent Set is used for the quantitative determination of Alanine Aminotransferase in serum.
- Catalyzes the transfer of the amino group from L-alanine to α-ketoglutarate
- Simultaneous oxidation of NADH to NAD
- Rate of decrease in absorbance is directly proportional to ALT activity
ALT Liquid Reagent Set is used for the quantitative determination of Alanine Aminotransferase in serum
The Olympus kit is for use on the Beckman Coulter AU 400, 480, and 640 analyzers and is for in vitro diagnostic use only. There are six 67 mL R1 Reagent bottles and five 17 mL R2 Reagent bottles in the Olympus kit.
Certifications: CE Compliant
Ordering information: Materials Required but not Provided:
The validity of the reaction should be monitored using control sera with known normal and abnormal ALT (SGPT) values (C7590/91/92).
The validity of the reaction should be monitored using control sera with known normal and abnormal ALT (SGPT) values (C7590/91/92).
Frequently Bought Together