About this item
These high-performance Endurosaf™ aprons feature 8- to 9-millimeter die-cut Enduro 2000™ polyurethane material that is strong and tough and offer users a unique combination of characteristics that outperform neoprene, nitrile, vinyl, and other films.
- Sealed ties provide a durable, consistent fit
- Remain flexible in cold temperatures
- Strong and tough fabrication
Sealed ties and adjustable neck straps are strong and provide a durable, consistent fit for a wide variety of users
Wherever oil, grease and chemicals are present, Ansell aprons help workers keep clean and protected from harmful contact. Endurosaf™ aprons comply with FDA regulations for food contact and are exceptionally resistant to abrasion, chemicals, oils and fats.
Neck strap features an adjustable non-corrosive, non-absorbent plastic buckle for a more comfortable fit. 56-800 series aprons are soft, comfortable, easy to clean and remain flexible in cold temperatures. Use in the chemical processing, food processing, janitorial/sanitation, medical laboratories, pharmaceutical, and research laboratory industries. Uses include medical, cleanrooms, laboratories, food processing, chemical processing, pharmaceutical, and janitorial.
Specifications
- Catalog No:
- 32891-452
- 32891-450
- Model:
- 56-800
- 56-800
- Garment application:
- Food, Industrial, Cleanroom, Chemical protection, Laboratory, Pharmaceutical
- Food, Industrial, Cleanroom, Chemical protection, Laboratory, Pharmaceutical
- Risk area:
- Cuts
- Cuts
- Gender:
- Unisex
- Unisex
- Fabric:
- Polyurethane
- Polyurethane
- Thickness:
- 8 - 9 mil
- 8 - 9 mil