Gemini ESD Mats, Bulk Rolls, Light Gray, ACL
Supplier: Avantor
Certificates
About this item
Gemini ESD Mats are dual-layer, static dissipative, polymer-based materials made for bench tops and grounded workbench surfaces
Bulk roll mats are provided in 50' lengths, and are available in 24", 30", and 36" widths. They are 100% recyclable, and made in the USA.
Mats offer a lifetime warranty on electrical resistance (10⁶ to 10⁸ ohms).