About this item
PLASMID+® liquid media supports much higher cell densities and plasmid yields than LB media
Optimal shake flask yields are achieved using Ultra Yield™ Flasks, which facilitate maximum culture aeration. PLASMID+® media may also be used in a bioreactor with continuous aeration and agitation. E. coli DH5α is the preferred host strain for use with PLASMID+®. E. coli XL1-Blue also produces high quality plasmid DNA and may improve plasmid DNA yields with plasmids smaller than 3kb.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Sterility:Sterile