Control Serum, Wako
About this item
Wako Control Serum is for research use only and is not for use in diagnostic procedures.
- For research use only
- Not for use in diagnostic procedures
- Each Control Serum is intercompatible
- Get five bottles of Serum I and Serum II at 5 mL each in the Combo pack
Contains five bottles of Control Serum at 5 mL each
The Control Serum Combo contains five bottles of Control Serum I and five bottles of Control Serum II at 5mL each
For research use only; Not for use in diagnostic procedures.
Ordering information: Each Control Serum is intercompatible. Also compatible with HDL-C/LDL-C Calibrator, HDL Cholesterol E, HDL-C Reagent 1 L-Type, HDL-C Reagent 2 L-Type, LDL-C Reagent 1 L-Type, LDL-C Reagent 2 L-Type, NEFA Linearity Set, NEFA Standard Solution, NEFA-HR2 Color A, NEFA-HR2 Color B, NEFA-HR2 Solvent A, NEFA-HR2 Solvent B, Triglyceride M Enzyme Color A L-Type, and Triglyceride M Enzyme Color B L-Type
3 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Wako Control Serum is for research use only and is not for use in diagnostic procedures.
- For research use only
- Not for use in diagnostic procedures
- Each Control Serum is intercompatible
- Get five bottles of Serum I and Serum II at 5 mL each in the Combo pack
Contains five bottles of Control Serum at 5 mL each
The Control Serum Combo contains five bottles of Control Serum I and five bottles of Control Serum II at 5mL each
For research use only; Not for use in diagnostic procedures.
Ordering information: Each Control Serum is intercompatible. Also compatible with HDL-C/LDL-C Calibrator, HDL Cholesterol E, HDL-C Reagent 1 L-Type, HDL-C Reagent 2 L-Type, LDL-C Reagent 1 L-Type, LDL-C Reagent 2 L-Type, NEFA Linearity Set, NEFA Standard Solution, NEFA-HR2 Color A, NEFA-HR2 Color B, NEFA-HR2 Solvent A, NEFA-HR2 Solvent B, Triglyceride M Enzyme Color A L-Type, and Triglyceride M Enzyme Color B L-Type
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 80076-216
- 80076-218
- 80076-220
- Stability:
- Until expiration or seven days after reconstitution.
- Storage Temperature:
- 2 - 10° C
- 2 - 10° C
- 2 - 10° C
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- 80076-216Control Serum ISupplier Number: 410-00102Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.80076-216StabilityStorage Temperature2 - 10° C - 80076-218Control Serum IISupplier Number: 416-00202Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.80076-218StabilityStorage Temperature2 - 10° C - 80076-220Control Serum ComboSupplier Number: 416-00102Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.80076-220StabilityUntil expiration or seven days after reconstitution.Storage Temperature2 - 10° C
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



