Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G
- Antigen Symbol:IgG
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Flow Cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Size:1.5 mg
- Epitope:F(c) γ
- Cross Adsorption:No
- Form:Freeze-dried powder
- Storage Buffer:0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6, 15mg/mL BSA, 0.05% Azide
- Storage Temperature:+4 °C
- Concentration:1.0-1.5 mg/mL
- Shipping Temperature:ambient
- Purification:Affinity Purified
- Cat. No.:102645-978
- Packaging:Glass ampoule
- Supplier no.:112-605-008
Specifications
About this item
Whole IgG antibodies are isolated as intact molecules from antisera by immunoaffinity chromatography. They have an Fc portion and two antigen binding Fab portions joined together by disulfide bonds and therefore they are divalent. The average molecular weight is reported to be about 160 kDa. The whole IgG form of antibodies is suitable for the majority of immunodetection procedures and is the most cost effective. Based on immunoelectrophoresis and/or ELISA, the antibody reacts with the Fc portion of rat IgG heavy chain but not with the Fab portion of rat immunoglobulins. No antibody was detected against rat IgM or against non-immunoglobulin serum proteins. The antibody may cross-react with immunoglobulins from other species.
Type:
Antigen: IgG Fcγ
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat